missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PTRF Polyclonal antibody specifically detects PTRF in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | PTRF |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CAVIN, Cavin-1, CGL4, FLJ90031, polymerase I and transcript release factor, RNA polymerase I and transcript release factor, TTF-I interacting peptide 12 |
| Host Species | Rabbit |
| Immunogen | PTRF antibody was raised against a 19 amino acid peptide near the carboxy terminus of human PTRF. The immunogen is located within amino acids 270 - 320 of PTRF. |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?