missing translation for 'onlineSavingsMsg'
Learn More

PTPRH Antibody, Novus Biologicals™

Product Code. 18126601 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18126601 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18126601 Supplier Novus Biologicals Supplier No. NBP220005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTPRH Polyclonal specifically detects PTPRH in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRH
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9HD43
Gene Alias EC 3.1.3.48, FLJ39938, MGC133058, MGC133059, protein tyrosine phosphatase, receptor type, H, receptor-type tyrosine-protein phosphatase H, R-PTP-H, SAP1, SAP-1, Stomach cancer-associated protein tyrosine phosphatase 1, Transmembrane-type protein-tyrosine phosphatase type H
Gene Symbols PTPRH
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human PTPRH. The exact sequence is proprietary.
Molecular Weight of Antigen 122 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5794
Test Specificity Human. Species reactivity based on sequence identity: Mouse (80%), Rat (82%).
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.