Learn More
Bohlender™ PTFE US Spherical Ground Joint Sleeves
Provides for high-vacuum tight sealing with a low leak rate.
Brand: Bohlender™ H931-04
Additional Details : Weight : 0.60000kg
Description
- Providing temperature resistance from −200° to +250°C
- Universal chemical resistance
- FDA conformity
- Vacuum suitable
- Reusable
- Made of PTFE with gripping ring and sealing rings on the outside
- Low leak rate of less than 1 x 0.01 mbar
- Solid construction for continued use
- For US standard size 18 KS
Specifications
Providing high-vacuum seals | |
Spherical ground joint sleeve | |
−200° to +250°C |
US standard 18 KS | |
PTFE |