Learn More
Masterflex™ PTFE Tubing, 250 ft. spool
Product Code. 11760139
Click to view available options
Diameter (Metric) Inner:
1.6 mm
6.4 mm
Unit Size:
Each
Product Code. 11760139
Brand: Masterflex™ 9523100
Only null left
Add to basket
Add to basket
Ideal for demanding lab and chemical applications
  • PTFE tubing is extremely chemically resistant.
  • Available in spooled rolls for increased versatility.
  • Suitable for use in a broad range of temperatures, from -400 to +500°F (-240 to +260°C).
  • Resin is FDA-compliant (21 CFR 177.1550) for high-purity work.
  • Translucent.
  • Sterilize via autoclave.
Specifications
Certifications/Compliance 3A;FDA;NSF-61;NSF-51;ADCF;Non-DEHP;European Pharmacopoeia;Non-Phthalate;USP
Autoclavable Autoclavable
Diameter (Metric) Inner 1.6 mm
Material PTFE
Diameter (Metric) Outer 3.2 mm
Overall Length (English) 250 ft.
Overall Length (Metric) 76.2 m
Thickness (Metric) 0.8 mm
Packaging Quantity 250 ft./Pk.
Sterilization Method Autoclave, Ethylene Oxide or Gamma
Color Translucent