Learn More
Hirschmann™ PTFE-Stopcock with PP Tip
Product Code. 10263721
Change view
Click to view available options
Material:
PTFE
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
10263721 PTFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10263721 Supplier Hirschmann™ Supplier No. 8901076
Only null left
Add to basket
Add to basket
Component for burette assembly with burette tube and seperate PTFE stopcock with PP tip.
  • Interchangeable
  • To be used with burette tubes no. 307 12
Specifications
Type PTFE to stopcock with PP tip
Description Interchangeable, can be fitted to burette tube 307 12 60 - 307 12 75
Material PTFE