Learn More
VITLAB™ PTFE Polygonal Magnetic Stirring Bar
Product Code. p-8004642
Click to view available options
Length (Metric):
5 mm
8 mm
10 mm
12 mm
15 mm
20 mm
25 mm
30 mm
40 mm
60 mm
70 mm
80 mm
Diameter (Metric):
2 mm
3 mm
4.5 mm
6 mm
7 mm
8 mm
10 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 16513602
Brand: VITLAB™ 301097
Only null left
Add to basket
Add to basket
Ensure long-lasting durability and prevent demagnetization of Vitlab™ PFTE Polygonal Magnetic Stirring Bars via proper storage and not dropping them on hard surfaces (e.g. steel).
  • Permanent magnet AlNiCo V core
  • Angled shape gives rise to significant turbulence
  • Achieves effective mixing, even at low spin rates
  • FTE
Specifications
For Use With (Application) For preparation, separation and concentration
Includes Permanent magnet AlNiCo V core
Diameter (Metric) 3 mm
Length (Metric) 10 mm
Material PTFE
Quantity 5/Pk.
Shape Polygon