Learn More
VITLAB™ PTFE Polygonal Magnetic Stirring Bar
Click to view available options
Length (Metric):
5 mm
8 mm
10 mm
12 mm
15 mm
20 mm
25 mm
30 mm
40 mm
60 mm
70 mm
80 mm
Diameter (Metric):
2 mm
3 mm
4.5 mm
6 mm
7 mm
8 mm
10 mm
Pakningsstørrelse:
Pack of 10
Pack of 5
Beskrivelse
- Permanent magnet AlNiCo V core
- Angled shape gives rise to significant turbulence
- Achieves effective mixing, even at low spin rates
- FTE
Tekniske data
Tekniske data
| For Use With (Application) | For preparation, separation and concentration |
| Includes | Permanent magnet AlNiCo V core |
| Diameter (Metric) | 3 mm |
| Length (Metric) | 10 mm |
| Material | PTFE |
| Quantity | 5/Pk. |
| Shape | Polygon |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion