Description
- Safe and chemically resistant
- For reasons of work safety, please ensure that the fittings with the correct diameter are used
Specifications
Specifications
| Diameter (Metric) Inner | 4.75 mm |
| Length (Metric) | 3 m |
| For Use With (Equipment) | Safety Caps and Fittings |
| Material | PTFE |
| For Use With (Application) | HPLC Systems |
| Type | Capillary |
| Diameter (Metric) Outer | 6.35 mm |