Learn More
Bohlender™ PTFE Blunt Tip Tweezers
Product Code. p-8557032
Click to view available options
Length (Metric):
150 mm
200 mm
Unit Size:
Each
Product Code. 10736182
Brand: Bohlender™ H91206
Only null left
Add to basket
Add to basket
Use these tweezers for a variety of applications. BOLA PTFE Tweezers feature universal chemical resistance.
  • Precast tweezers
  • PTFE construction
  • With pointed or blunt ends
  • Universal chemical resistance
Specifications
Tip Style Blunt
No. per Pack Each
Design Precast tweezer
Material PTFE
Length (Metric) 200 mm
Product Type Tweezers