Learn More
Pterostilbene, MedChemExpress
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Pterostilbene is a stilbenoid isolated from blueberries and Pterocarpus marsupium. Shows anti-oxidant, anti-inflammatory, anti-carcinogenic, anti-diabetic and anti-obesity properties. Pterostilbene blocks ROS production, also exhibits inhibitory activity against various free radicals such as DPPH, ABTS, hydroxyl, superoxide and hydrogen peroxide.
Specifications
Specifications
| Chemical Name or Material | Pterostilbene |
| CAS | 537-42-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H16O3 |
| Quantity | 25 mg |
| Solubility Information | DMSO : 110 mg/mL (429.18 mM; Need ultrasonic) |
| SMILES | OC1=CC=C(/C=C/C2=CC(OC)=CC(OC)=C2)C=C1 |
| Molecular Weight (g/mol) | 256.3 |
| Formula Weight | 256.3 |
| Show More |