missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTCD3 Monoclonal Antibody (OTI1A8), TrueMAB™, OriGene
Description
PTCD3 Monoclonal antibody specifically detects PTCD3 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PTCD3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1A8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | Mlh1 |
| Gene Accession No. | P40692 |
| Gene Alias | 2610034F17Rik; 2810422B04Rik; 28S ribosomal protein S39, mitochondrial; AA589622; AU045708; Mitochondrial small ribosomal subunit protein mS39; Mrps39; MRP-S39; pentatricopeptide repeat domain 3; pentatricopeptide repeat domain-containing protein 3, mitochondrial; pentatricopeptide repeat-containing protein 3, mitochondrial; Ptcd3; similar to CG4679-PA; transformation-related gene 15 protein; TRG15; TRG-15 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?