missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSPH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14513-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to a subfamily of the phosphotransferases. This encoded enzyme is responsible for the third and last step in L-serine formation. It catalyzes magnesium-dependent hydrolysis of L-phosphoserine and is also involved in an exchange reaction between L-serine and L-phosphoserine. Deficiency of this protein is thought to be linked to Williams syndrome.Specifications
| PSPH | |
| Polyclonal | |
| Unconjugated | |
| PSPH | |
| L 3 phosphoserine phosphatase, phosphoserine phosphatase, PSP, PSPase, PSPH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100678, 304429, 5723 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P78330, Q5M819, Q99LS3 | |
| PSPH | |
| PSPH Fusion Protein Ag5972 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction