missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PSGR Monoclonal antibody specifically detects PSGR in Human samples. It is validated for Immunohistochemistry
Specifications
Specifications
| Antigen | PSGR |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 1033009 |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | HPRAJ, Olfactory receptor OR11-16, olfactory receptor, family 51, subfamily E, member 2, olfactory receptor, family 51, subfamily E, member 3 pseudogene, olfactory receptor, family 52, subfamily A, member 2, OR51E3P, OR52A2, prostate specific G-protein coupled receptor, Prostate-specific G-protein coupled receptor, PSGRolfactory receptor 51E2 |
| Host Species | Mouse |
| Immunogen | Human PSGR synthetic peptide, Accession # Q9H255 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?