missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587218
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
Human pregnancy-specific glycoproteins (PSG), are a group of closely related secreted glycoproteins which are highly expressed in placental syncytiotrophoblast cells of fetal origin. PSG are commonly expressed in tumors of trophoblast origin (hydatidiform mole, choriocarcinoma). They represent the most abundant fetal protein in the maternal blood at term. The eleven human PSG genes (PSG1-PSG11) form together with the carcinoembryonic antigen (CEA)-related cell adhesion molecule (CEACAM) genes the CEACAM gene family. PSG stimulates secretion of TH2-type cytokines from monocytes. CD9 was shown to represent the monocyte receptor for murine PSG17. PSG are thought to modulate the maternal immune system during pregnancy thus protecting the semiallotypic fetus from rejection.
Specifications
| PSG1 | |
| Polyclonal | |
| Unconjugated | |
| PSG1 | |
| B1G1; CD66 antigen-like family member F; CD66f; DHFRP2; Fetal liver non-specific cross-reactive antigen 1/2; FLJ90598; FLJ90654; FL-NCA-1/2; PBG1; pregnancy specific beta-1-glycoprotein 1; pregnancy-specific B-1 glycoprotein; pregnancy-specific beta-1 glycoprotein C/D; Pregnancy-specific beta-1-glycoprotein 1; PS-beta-C/D; PS-beta-G-1; PSBG1; PSBG-1; PSG 1; PSG IIA-d; PSG1; PSG-1d; PSG95; PSGGA; PSGIIA; PSGIIA-a; PSGIIA-b; PSGIIA-c; SP 1; SP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5669 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P11464 | |
| PSG1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 180-419 of human PSG1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction