missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PSEM 308 hydrochloride

Description
PSEM 308 hydrochloride is a PSAM (pharmacologically selective actuator module) agonist. Activates PSAML141F-GlyR chimeric ion channels. Inhibits activity of neurons expressing PSAML141F-GlyR in vivo and activates locus coeruleus noradrenergic neurons expressing PSAML141F,Y115F-5-HT3 ion channels.
Recommended concentration for use in mice is 5 mg/kg or lower.
Plasmid vectors for the transfection of cells with PSAML141F-GlyR and PSAML141F,Y115F-5-HT3 are available from Addgene.
Specifications
Specifications
| Chemical Name or Material | 5-Methyl-5,8,9,10,11 A,12-hexahydro-4 H-8,11-ethanopyrido[3 ',2 ':3,4 ]diazepino[6,7,1-hi]indole hydrochloride |
| Quantity | 25 mg |
| Target | Pharmacologically Selective Effector Molecules/PSEM |
| Molecular Formula | C17H21N3.HCl |
| Formula Weight | 303.83 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?