missing translation for 'onlineSavingsMsg'
Learn More

PRT062607 Hydrochloride, MedChemExpress

Product Code. 16419047
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16419047

Brand: Medchem Express HY15323

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PRT062607 Hydrochloride (P505-15 Hydrochloride) is a highly specific and potent inhibitor of purified Syk (IC50 1-2 nM).

PRT062607 Hydrochloride (P505-15 Hydrochloride) is a highly specific and potent inhibitor of purified Syk (IC50 1-2 nM).

Specifications

Chemical Name or Material PRT062607 Hydrochloride
CAS 1370261-97-4
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C19H24ClN9O
Quantity 5 mg
Synonym P505-15 Hydrochloride
Solubility Information H2O : ≥ 50 mg/mL (116.30 mM) ∣DMSO : ≥ 33 mg/mL (76.76 mM)
SMILES NC(C1=CN=C(N[C@H]2[C@@H](N)CCCC2)N=C1NC3=CC=CC(N4N=CC=N4)=C3)=O.[H]Cl
Molecular Weight (g/mol) 429.91
Formula Weight 429.91
Percent Purity 97.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.