missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRPH2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16642775 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16642775 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16642775 Supplier Invitrogen™ Supplier No. PA5106336

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PRPH2.

Peripherin-2 (PRPH2), also known as retinal degeneration slow protein (RDS), is a photoreceptor-specific tetraspanin protein implicated in outer segment disk morphogenesis. It may function as an adhesion molecule involved in stabilization and compaction of outer segment disks or in the maintenance of the curvature of the rim. Mutations in peripherin-2 are responsible for various retinal degenerative diseases including autosomal dominant retinitis pigmentosa (ADRP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPH2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRPH2
Gene Accession No. P15499, P17438, P23942
Gene Alias AOFMD; AVMD; CACD2; DS; MDBS1; Nmf193; peripherin 2; peripherin 2 (retinal degeneration, slow); peripherin 2, homolog of mouse; peripherin, photoreceptor type; peripherin-2; PRPH; PRPH2; Rd2; Rd-2; RDS; retinal degeneration 2; retinal degeneration slow protein; retinal degeneration, slow; retinal degeneration, slow (retinitis pigmentosa 7); retinal peripherin; RP7; RSRDS; tetraspanin-22; TSPAN22; Tspan-22
Gene Symbols PRPH2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRPH2(Accession P23942), corresponding to amino acid residues P147-K197.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19133, 25534, 5961
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.