Learn More
MedChemExpress Protirelin acetate
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Purity | 99.93% |
| CAS | 120876-23-5 |
| Content And Storage | Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| For Use With (Application) | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 452.46 g/mol |
| Product Type | Peptides |
| Quantity | 50 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 250 mg/mL (552.54 mM∣Need ultrasonic) ∣H2O : 50 mg/mL (110.51 mM∣Need ultrasonic) |
| Synonym | Thyrotropin-releasing-hormone acetate∣TRH acetate |