missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Protino Ni-IDA Resin for His-tag Protein Purification

Product Code. 11912292
Click to view available options
Quantity:
120 g
600 g
Unit Size:
120g
600g
This item is not returnable. View return policy

Product Code. 11912292

Brand: Macherey-Nagel™ 745210.120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The matrix of choice for highest protein purity as dry bulk material.

Protino™ Ni-IDA, the purer alternative for higher yields of His-tag proteins

  • Chelating group IDA, three protein binding sites
  • High protein yield/recovery even from diluted samples
  • High protein concentration
  • High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
  • Dry resin, storage at room temperature

Protino™ Ni-IDA packed columns

  • Ready to use pre-packed columns filled with dry Protino™ Ni-IDA resin
  • Buffers included

Protino™ Ni-IDA resin

  • Dry bulk matrix
  • Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink or swell - highest flexibility
  • Batch procedure, gravity flow column chromatography, FPLC™ applications
  • Empty Protino™ columns available for use with Protino™ Ni-IDA resin
  • Empty 96 well receiver plates available for use with Protino™ Ni-IDA resin

Protino™ Multi-96 Ni-IDA

  • Ready to use pre-packed 96 well plates filled with dry Protino™ Ni-IDA resin
  • Buffers included

For purification of polyhistidine-tagged proteins, batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.

Specifications

Bed Volume 10 mg/mL
Binding Capacity 20mg/g
Flow Rate 0.5 to 1.0 mL/min./cm2
Kit Contents Dry silica bulk resin
Matrix Macroporous silica
Content And Storage Room Temperature
Max. Operating Pressure 145 psi
Quantity 120 g
For Use With (Application) Purification of polyhistidine-tagged proteins
Type Resin
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1A
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P391-Collect spillage.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : resin with Ni in nickel(II) complexes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.