Learn More
Macherey-Nagel™ Protino Glutathione Agarose 4B for GST-tag Protein Purification
Product Code. 15810998
Click to view available options
Quantity:
1 mL
10 mL
100 mL
Unit Size:
100mL
10mL
1mL
Do not autoclave the gel.
Product Code. 15810998
Brand: Macherey-Nagel™ 745500.1
Only null left
Add to basket
Add to basket
Do not autoclave the gel.
Best choice for cost effective GST-tag protein purification.
Protino™ Glutathione Agarose 4B is an affinity chromatography medium that enables fast, single-step purification of glutathione-S-transferase (GST) fusion proteins and other glutathione-binding proteins. Purification of fusion proteins from a whole cell lysate is based on the strong affinity of the GST moiety for glutathione, which is immobilized on agarose beads.
  • Highest performance–Equivalent to Glutathione Sepharose™ 4B / GSTrap™ 4B columns
  • Simply replace your current products—No optimization or protocol change necessary
  • Universal usage—Suitable for small proteins, large protein complexes, proteins with low expression rates universal use
  • Highest flexibility of applications—For batch binding, gravity-flow column chromatography, MPLC/FPLC™
Specifications
Capacity (Metric) 1 mL
Description Glutathione Agarose
Quantity 1 mL
Type Agarose
Sample Volume 1 mL
compliance-icons
Product Identifier
  • Protino Glutathione Agarose 4B
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Supplemental information
  • MIXTURE LIST-Contains : ethanol (diluted < 20 %) (denatured with1%IPA/1%MEK, acc.2016/1867/EU)