missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 20S beta2 Antibody [DyLight 594], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Proteasome 20S beta2 Polyclonal antibody specifically detects Proteasome 20S beta2 in Human,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Proteasome 20S beta2 |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.4.25.1, HC7-I, Macropain subunit C7-I, MGC104215, MGC126885, multicatalytic endopeptidase complex subunit C7-1, Multicatalytic endopeptidase complex subunit C7-I, proteasome (prosome, macropain) subunit, beta type, 2, proteasome beta 2 subunit, Proteasome component C7-I, proteasome subunit beta type-2, proteasome subunit, beta type, 2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-201 of human Proteasome 20S beta2 (NP_002785.1).,, Sequence:, MEYLIGIQGPDYVLVASDRVAASNIVQMKDDHDKMFKMSEKILLLCVGEAGDTVQFAEYIQKNVQLYKMRNGYELSPTAAANFTRRNLADCLRSRTPYHVNLLLAGYDEHEGPALYYMDYLAALAKAPFAAHGYGAFLTLSILDRYYTPTISRERAVELLRKCLEELQKRFILNLPTFSVRIIDKNGIHDLDNISFPKQGS |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?