missing translation for 'onlineSavingsMsg'
Learn More

PROTAC BRD9-binding moiety 1 hydrochloride, MedChemExpress

Product Code. 16415106
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16415106 100 mg 100mg
16450487 50 mg 50mg
16442966 25 mg 25mg
16426837 5 mg 5mg
16414138 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16415106 Supplier Medchem Express Supplier No. HY107445A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PROTAC BRD9-binding moiety 1 hydrochloride is a compound that binds to BRD9, and used for inhibiting BRD9 activity, based on PROTAC.

PROTAC BRD9-binding moiety 1 hydrochloride is a compound that binds to BRD9, and used for inhibiting BRD9 activity, based on PROTAC.

Specifications

Chemical Name or Material PROTAC BRD9-binding moiety 1 hydrochloride
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H26ClN3O7S2
Quantity 100 mg
Solubility Information DMSO : 20.83 mg/mL (37.46 mM; Need ultrasonic) ∣H2O : 2.5 mg/mL (4.50 mM; Need ultrasonic)
SMILES O=C(O)COC1=CC=C(C2=CN(C)C(C3=C2SC(C(NC(CC4)CCS4(=O)=O)=N)=C3)=O)C=C1OC.Cl
Molecular Weight (g/mol) 556.05
Formula Weight 556.05
Percent Purity 97.07%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.