missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Prostate Specific Antigen (PSA), Rabbit Monoclonal Antibody

Employ part of a comprehensive antibody portfolio Epredia™ Prostate Specific Antigen (PSA), Rabbit Monoclonal Antibody. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Brand:  Epredia™ RM-2104-S0

 View more versions of this product

Product Code. 12653617

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

PSA is a chymotrypsin-like serine protease (kallikrein family) produced by the prostate epithelium, and abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. PSA is used to confirm prostatic acinar cell origin in primary and metastatic carcinoma and to rule out non-prostatic carcinoma mimics.

Host Species: Rabbit

Clone: EP1588Y

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues near the C-terminus of human PSA

Molecular Weight: 34kDa

Positive Control: Normal prostate or prostate carcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Prostate Specific Antigen (PSA)
Monoclonal
Unconjugated
A synthetic peptide corresponding to residues near the C-terminus of human PSA
ASR
Primary
Concentrated
Immunohistochemistry (Paraffin)
EP1588Y
Rabbit
100 μL
Cancer and Tumor Biology
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.