missing translation for 'onlineSavingsMsg'
Learn More

Prostaglandin E Synthase Antibody, Novus Biologicals™

Product Code. 18628885 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18628885 100 μg 100µL
18608985 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18628885 Supplier Novus Biologicals Supplier No. NBP299619100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Prostaglandin E Synthase Polyclonal antibody specifically detects Prostaglandin E Synthase in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prostaglandin E Synthase
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:500-1:2000
Formulation PBS (pH 7.0)
Gene Alias EC 5.3.99.3, MGC10317, MGST1L1MGST1-like 1, MGST1-L1mPGES-1, MGST-IV, Microsomal glutathione S-transferase 1-like 1, Microsomal prostaglandin E synthase 1, MPGES, MPGES1, MPGES-1, p53-induced apoptosis protein 12, p53-induced gene 12 protein, PGESmicrosomal prostaglandin E synthase-1, PIG12glutathione S-transferase 1-like 1, PP102, PP1294, prostaglandin E synthase, TP53I12, tumor protein p53 inducible protein 12
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Prostaglandin E Synthase.
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9536
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.