Learn More
DiscoverX Propranolol Hydrochloride
Click to view available options
:
10 mg
Description
Propranolol is a sympatholytic beta-adrenergic antagonist, widely used for treatment of tremors, angina, hypertension, heart rhythm disorders, migraines, thyrotoxicosis, anxiety, and other circulatory diseases. It was the first successful beta-blocker developed. Acts as an antagonist for ADRAB1 and ADRAB2. Provided as 10mg of dry powder.
Specifications
Specifications
| Quantity | 10 mg |