Learn More
Bochem™ Propeller Stirrer, 4 Vertical Blades
Product Code. 10739271
Click to view available options
For Use With (Equipment):
Centrifuge
Unit Size:
Each
Product Code. 10739271
Brand: Bochem™ 9056
Please to purchase this item. Need a web account? Register with us today!
Mix media with low and middle viscosity. Bochem™ Propeller Stirrer, 4 Vertical Blades is constructed of 18/10 stainless steel.
  • 18/10 stainless steel construction.
  • Middle RPM range.
  • Mixes media with low and middle viscosity.
Specifications
Type Vertical Blade
Material Stainless Steel (18/10)
For Use With (Equipment) Centrifuge
Length (Metric) 300 mm
Width (Metric) 100 mm