Learn More
Bochem™ Propane Bunsen Burners
Product Code. 10525031
Click to view available options
Diameter (Metric):
76 mm
80 mm
Unit Size:
Each
Product Code. 10525031
Brand: Bochem™ 7010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Bunsen Burner
Gas Type Natural Gas, Propane
Material Nickel, Steel
Diameter (Metric) 76 mm
Height (Metric) 165 mm
Max. Temperature (Metric) 1300°C
Weight (Metric) 0.25 kg