missing translation for 'onlineSavingsMsg'
Learn More

Promethazine hydrochloride, MedChemExpress

Product Code. 16471058
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16471058

Brand: Medchem Express HYB0781

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Promethazine hydrochloride is the first-generation antihistamine; strong antagonist of the H1 receptor and moderate mACh receptor antagonist, moderate affinity for 5-HT2A, 5-HT2C, D2 and α1-adrenergic receptors.

Promethazine hydrochloride is the first-generation antihistamine

Specifications

Chemical Name or Material Promethazine hydrochloride
Physical Form Solid
CAS 58-33-3
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C17H21ClN2S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : 100 mg/mL (311.64 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (155.82 mM; Need ultrasonic)
SMILES CC(N(C)C)CN1C(C=CC=C2)=C2SC3=CC=CC=C13.[H]Cl
Molecular Weight (g/mol) 320.88
Formula Weight 320.88
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.