missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Prolyl endopeptidase-like 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Cell Biology |
| Target | Prolyl endopeptidase-like |
| Synonym | EC 3.4.21, EC 3.4.21.-, EC 3.4.21.83, KIAA0436FLJ16627, prolyl endopeptidase-like, Prolylendopeptidase-like, putative prolyl oligopeptidase |
| Gene Symbol | PREPL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction