missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolactin R Antibody (1D3), Novus Biologicals™
Human Monoclonal Antibody
343.00 € - 533.00 €
Specifications
| Antigen | Prolactin R |
|---|---|
| Clone | 1D3 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30035661
|
Novus Biologicals
NBP3-26368-50ul |
50 μL |
343.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30036248
|
Novus Biologicals
NBP3-26368-100ul |
100 μL |
533.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prolactin R Monoclonal antibody specifically detects Prolactin R in Human, Mouse samples. It is validated for ELISASpecifications
| Prolactin R | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein | |
| Recombinant Human Prolactin R protein [UniProt P16471] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 1D3 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Apoptosis | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 5618 | |
| IgG2 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title