Learn More
Prohydrojasmon racemate, MedChemExpress
Product Code. 16475416
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16475416
Brand: Medchem Express HY112101
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Prohydrojasmon racemate (n-Propyl dihydrojasmonate) is the racemate of Prohydrojasmon. Prohydrojasmon is a synthesized plant growth regulator.
Prohydrojasmon racemate (n-Propyl dihydrojasmonate) is the racemate of Prohydrojasmon. Prohydrojasmon is a synthesized plant growth regulator.
Specifications
Chemical Name or Material Prohydrojasmon racemate
Physical Form Liquid
CAS 158474-72-7
Color Colorless
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H26O3
Synonym n-Propyl dihydrojasmonate
Solubility Information DMSO : ≥ 54 mg/mL (212.29 mM)
SMILES O=C(OCCC)CC1C(CCCCC)C(CC1)=O
Molecular Weight (g/mol) 254.37
Formula Weight 254.37
Percent Purity 98.0%
Grade Research