missing translation for 'onlineSavingsMsg'
Learn More

Proenkephalin-A Rabbit anti-Human, Polyclonal, Proteintech

Product Code. p-7204568 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16821794 20 μL 20µL
16811794 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16821794 Supplier Proteintech Supplier No. 135311AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

neuropeptide hormone; plays a role in the responses to stress and pain perception.

Specifications

Antigen Proenkephalin-A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PENK
Gene Accession No. P01210
Gene Alias Opioid growth factor, PENK, proenkephalin, Proenkephalin A
Gene Symbols PENK
Host Species Rabbit
Immunogen Proenkephalin-A Fusion Protein Ag4428
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5179
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.