missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PROCR Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00 € - 349.00 €
Specifications
| Antigen | PROCR |
|---|---|
| Clone | 1F5C4 |
| Concentration | 1273 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234832
|
Proteintech
67658-1-IG-150UL |
150 μL |
410.00 € 349.00 € / 150µL Save 61.00 € 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236292
|
Proteintech
67658-1-IG-20UL |
20 μL |
145.00 € 129.00 € / 20µL Save 16.00 € 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PROCR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PROCR. This antibody recognizes human, pig antigen. The PROCR antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| PROCR | |
| 1273 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 10544 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1F5C4 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Pig | |
| CCCA; CCD41; CD201; EPCR; MGC23024; bA42O4.2 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title