Learn More
Procinolol, TRC
Specifications
Specifications
| Chemical Name or Material | Procinolol |
| CAS | 27325-36-6 |
| Molecular Formula | C15H23NO2 |
| Quantity | 25 mg |
| Synonym | 1-(O-Cyclopropylphenoxy)-3-(isopropylamino)-2-propanol, 1-(2-Cyclopropylphenoxy)-3-[(1-methylethyl)amino]-2-propanol, (±)-Procinolol, SD 2124-01 |
| InChI Formula | InChI=1S/C15H23NO2/c1-11(2)16-9-13(17)10-18-15-6-4-3-5-14(15)12-7-8-12/h3-6,11-13,16-17H,7-10H2,1-2H3 |
| SMILES | CC(C)NCC(O)COc1ccccc1C2CC2 |
| IUPAC Name | 1-(2-cyclopropylphenoxy)-3-(propan-2-ylamino)propan-2-ol |
| Molecular Weight (g/mol) | 249.35 |
| Formula Weight | 249.1729 |