Learn More
(±)-Procinolol Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (±)-Procinolol Hydrochloride |
| CAS | 27325-18-4 |
| Molecular Formula | C15H24ClNO2 |
| Quantity | 10 mg |
| Synonym | 1-(2-Cyclopropylphenoxy)-3-[(1-methylethyl)amino]-2-propanol Hydrochloride, 1-(o-Cyclopropylphenoxy)-3-(isopropylamino)-2-propanol Hydrochloride, 1-(o-Cyclopropylphenoxy)-3-isopropylamino-2-propanol hydrochloride, NSC 303852 |
| InChI Formula | InChI=1S/C15H23NO2.ClH/c1-11(2)16-9-13(17)10-18-15-6-4-3-5-14(15)12-7-8-12;/h3-6,11-13,16-17H,7-10H2,1-2H3;1H |
| SMILES | OC(CNC(C)[C])COC1=C(C2CC2)C=CC=C1.Cl |
| Molecular Weight (g/mol) | 285.81 |