missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human Th9/Th17/Th22/Treg & Cytokine Panel 1C (14 plex)
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
Includes
- Human Standard Mix A
- Human Standard Mix B
- Human Bead Mix C
- Human Bead Mix D
- Human Detection Antibody Mix C
- Human Detection Antibody Mix D
- Reading Buffer
- Wash Buffer 10x
- Streptavidin-PE
- Universal Assay Buffer
- Detection Antibody Diluent
- PCR 8-Tube Strip
- Adhesive Film
- Greiner Microplate black 96 well
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
- This kit does not contain a Multiscreen Filter Plate
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Human |
| For Use With (Application) | Multiplex Immunoassay |
| Protein Subtype | IFN alpha, IL-1 alpha, IL-1 Receptor Antagonist, IL-10, IL-15, IL-17A, IL-21, IL-22, IL-23, IL-27, IL-31, IL-7, IL-9, TNF beta |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Sample Volume | Serum, Plasma: 25 μL; CCS: 50 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction