Learn More
Techne™ Probe Insert Block
Product Code. 15513670
Change view
Click to view available options
Diameter (Metric):
19.5 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
15513670 19.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15513670 Supplier Techne™ Supplier No. 1611547
Please to purchase this item. Need a web account? Register with us today!
Techne™ Probe Insert Block for use with Tecal™ High Temperature Dri-Block Calibrator
  • Change capacity, testing type, accuracy, or portability of Techne Calibration calibrators with these additional parts
  • For use with Techne High Temperature Dri-Block Calibrator
Specifications
For Use With (Equipment) Tecal™ High-Temperature Dri-Block Calibrator (650F/S/H)
Material Aluminum Bronze
Type Probe Insert
Diameter (Metric) 19.5 mm