missing translation for 'onlineSavingsMsg'
Learn More

PRN694, MedChemExpress

Product Code. p-200101714
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16445288 10 mg 10mg
16441279 1 mg 1mg
16413326 25 mg 25mg
16411497 50 mg 50mg
16408007 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16445288 Supplier Medchem Express Supplier No. HY12680
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PRN694 is an irreversible, highly selective and potent covalent interleukin-2-inducible T-cell kinase (ITK) and resting lymphocyte kinase (RLK) dual inhibitor with IC50s of 0.3 nM and 1.4 nM, respectively. PRN694 exhibits extended target residence time on ITK and RLK, enabling durable attenuation of effector cells in vitro and in vivo.

PRN694 is an irreversible, highly selective and potent covalent interleukin-2-inducible T-cell kinase (ITK) and resting lymphocyte kinase (RLK) dual inhibitor with IC50s of 0.3 nM and 1.4 nM, respectively. PRN694 exhibits extended target residence time on ITK and RLK, enabling durable attenuation of effector cells in vitro and in vivo.

Specifications

Chemical Name or Material PRN694
CAS 1575818-46-0
Color Light Brown
Purity Grade Notes Research
Molecular Formula C28H35F2N5O2S
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (229.92 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC=C(C(F)F)S1)NC2=NC3=CC(CN[C@@H](C)C(C)(C)C)=CC=C3N2C[C@@H]4N(C(C=C)=O)CCC4
Molecular Weight (g/mol) 543.67
Formula Weight 543.67
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.