Learn More
PRN694, MedChemExpress
Product Code. 16445288
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16445288 10 mg 10mg
16441279 1 mg 1mg
16413326 25 mg 25mg
16411497 50 mg 50mg
16408007 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16445288 Supplier Medchem Express Supplier No. HY12680
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress PRN694 is an irreversible, highly selective and potent covalent interleukin-2-inducible T-cell kinase (ITK) and resting lymphocyte kinase (RLK) dual inhibitor with IC50s of 0.3 nM and 1.4 nM, respectively. PRN694 exhibits extended target residence time on ITK and RLK, enabling durable attenuation of effector cells in vitro and in vivo.
PRN694 is an irreversible, highly selective and potent covalent interleukin-2-inducible T-cell kinase (ITK) and resting lymphocyte kinase (RLK) dual inhibitor with IC50s of 0.3 nM and 1.4 nM, respectively. PRN694 exhibits extended target residence time on ITK and RLK, enabling durable attenuation of effector cells in vitro and in vivo.
Specifications
Chemical Name or Material PRN694
CAS 1575818-46-0
Color Light Brown
Purity Grade Notes Research
Molecular Formula C28H35F2N5O2S
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (229.92 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC=C(C(F)F)S1)NC2=NC3=CC(CN[C@@H](C)C(C)(C)C)=CC=C3N2C[C@@H]4N(C(C=C)=O)CCC4
Molecular Weight (g/mol) 543.67
Formula Weight 543.67
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation