Learn More
PRN1371, MedChemExpress
Product Code. 16494666
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16494666
Brand: Medchem Express HY101768
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress PRN1371 is a highly selective and potent FGFR1-4 and CSF1R inhibitor with IC50s of 0.6, 1.3, 4.1, 19.3 and 8.1 nM for FGFR1, FGFR2, FGFR3, FGFR4 and CSF1R, respectively.
PRN1371 is a highly selective and potent FGFR1-4 and CSF1R inhibitor with IC50s of 0.6, 1.3, 4.1, 19.3 and 8.1 nM for FGFR1, FGFR2, FGFR3, FGFR4 and CSF1R, respectively.
Specifications
Chemical Name or Material PRN1371
CAS 1802929-43-6
Color White
Purity Grade Notes Research
Molecular Formula C26H30Cl2N6O4
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (44.53 mM; Need ultrasonic)
SMILES O=C1C(C2=C(Cl)C(OC)=CC(OC)=C2Cl)=CC3=CN=C(NC)N=C3N1CCCN4CCN(C(C=C)=O)CC4
Molecular Weight (g/mol) 561.46
Formula Weight 561.46
Percent Purity 99.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease