missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT6 Antibody (JE36-57), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32854
This item is not returnable.
View return policy
Description
PRMT6 Monoclonal antibody specifically detects PRMT6 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| PRMT6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| EC 2.1.1.-, EC 2.1.1.125, FLJ10559, FLJ51477, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6, Histone-arginine N-methyltransferase PRMT6, HMT1 hnRNP methyltransferase-like 6, HMT1 hnRNP methyltransferase-like 6 (S. cerevisiae), HRMT1L6, protein arginine methyltransferase 6, protein arginine N-methyltransferase 6 | |
| Synthetic peptide within Human PRMT6 aa 191-240/ 375. (Uniprot: Q96LA8) | |
| 100 μL | |
| Chromatin Research, Signal Transduction | |
| 55170 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE36-57 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction