missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRL1 Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
The protein encoded by this gene belongs to a small class of prenylated protein tyrosine phosphatases (PTPs), which contains a PTP domain and a characteristic C-terminal prenylation motif. PTPs are cell signaling molecules that play regulatory roles in a variety of cellular processes. This tyrosine phosphatase is a nuclear protein, but may primarily associate with plasma membrane. The surface membrane association of this protein depends on its C-terminal prenylation. Overexpression of this gene in mammalian cells conferred a transformed phenotype, which implicated its role in the tumorigenesis. Studies in rat suggested that this gene may be an immediate-early gene in mitogen-stimulated cells. PRL phosphatases increase cell proliferation by stimulating progression from G1 into S phase. PRL-1 function is regulated in a cell cycle-dependent manner and implicate PRL-1 in regulating progression through mitosis, possibly by modulating spindle dynamics.
Specifications
Specifications
| Antigen | PRL1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | PTP4A1 |
| Gene Accession No. | Q63739, Q93096 |
| Gene Alias | AA415290; AU019864; C130021B01; DKFZp779M0721; HH72; PRL1; Prl-1; protein tyrosine phosphatase 4a1; protein tyrosine phosphatase type IVA 1; protein tyrosine phosphatase type IVA protein 1; protein tyrosine phosphatase type IVA, member 1; protein-tyrosine phosphatase 4a1; Protein-tyrosine phosphatase of regenerating liver 1; PTP(CAAX1); PTP(CAAXI); PTP4A1; PTP4A1 protein; PTP4A2; PTPCAAX1; RP5-1148A21.2 |
| Gene Symbols | PTP4A1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?