missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRKCB2 (PKC beta II), GST Tag Recombinant Protein

Product Code. 18715455
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
This item is not returnable. View return policy

Product Code. 18715455

Brand: Invitrogen™ A41909

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length, Active The gene accession number for PRKCB2 (PKC beta II) is NP_002729.2.

The PKC family of serine/threonine kinases, including PRKCB2 (PKC beta II), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II, and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. Protein kinase C (PKC) can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. This protein kinase has been reported to be involved in many different cellular functions, such as B cell activation, apoptosis induction, endothelial cell proliferation, and intestinal sugar absorption. Studies in mice also suggest that this kinase may also regulate neuronal functions and correlate fear-induced conflict behavior after stress. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05771
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 15mM glutathione, 100mM NaCl, 20% glycerol, 5mM DTT and no preservative; pH 8
Gene ID (Entrez) 5579
Molecular Weight (g/mol) 103.7 kDa
Name Human PRKCB2 (PKC beta II), GST Tag
pH Range 8
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias A130082F03Rik; PKCB; PKC-B; PKC-beta; Prkcb; PRKCB1; Prkcb2; protein kinase C beta; protein kinase C beta I; protein kinase C beta II; protein kinase C beta type; protein kinase C beta-II; protein kinase C, beta; protein kinase C, beta 1; protein kinase C, beta 1 polypeptide
Common Name PKC beta-2
Gene Symbol PRKCB
KinaseFamily PKC Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Active
Form Liquid
Protein Subtype Serine⁄Threonine Kinases
Research Category Drug Development
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.