Learn More
PRKCB Mouse, anti-Human, Clone: 4B7A1, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | PRKCB |
|---|---|
| Clone | 4B7A1 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18058185
|
Proteintech
68620-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18076385
|
Proteintech
68620-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PRKCB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRKCB. This antibody recognizes human, pig, rabbit, rat, mouse antigen. The PRKCB antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PRKCB | |
| 500 μg/mL | |
| Liquid | |
| Human, Pig, Rabbit, Rat, Mouse | |
| PKC B, PKC beta, PKCB, PRKCB, PRKCB1, PRKCB2, Protein kinase C beta type, protein kinase C, beta | |
| Primary |
| 4B7A1 | |
| Western Blot, ELISA | |
| Mouse | |
| 5579 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.