missing translation for 'onlineSavingsMsg'
Learn More

Pritelivir, MedChemExpress

Product Code. 16436726
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436726

Brand: Medchem Express HY15303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pritelivir (AIC316), an inhibitor of the viral helicase-primase complex, exhibits antiviral activity in vitro and in animal models of herpes simplex virus (HSV) infection. Pritelivir is active against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2) with the IC50 of 0.02 μM against HSV1-2.

Pritelivir (AIC316), an inhibitor of the viral helicase-primase complex, exhibits antiviral activity in vitro and in animal models of herpes simplex virus (HSV) infection. Pritelivir is active against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2) with the IC50 of 0.02 μM against HSV1-2.

Specifications

Chemical Name or Material Pritelivir
Physical Form Powder
CAS 348086-71-5
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H18N4O3S2
For Use With (Application) COVID-19-anti-virus
Synonym AIC316 BAY 57-1293
Solubility Information DMSO : ≥ 33 mg/mL (81.99 mM)
SMILES O=C(N(C1=NC(C)=C(S(=O)(N)=O)S1)C)CC2=CC=C(C3=NC=CC=C3)C=C2
Molecular Weight (g/mol) 402.49
Formula Weight 402.49
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.