missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pristinamycin IA, MedChemExpress
Description
Pristinamycin IA (Mikamycin B) is a cycle-peptidic macrolactone antibiotic. Pristinamycin IA is a substrate of P-glycoprotein and inhibits its function. Pristinamycin IA is active against StaphyloEoccus and Srreptococcus.
Specifications
Specifications
| Chemical Name or Material | Pristinamycin IA |
| Physical Form | Powder |
| CAS | 3131-03-1 |
| Color | Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C45H54N8O10 |
| Synonym | Mikamycin B Mikamycin IA |
| Solubility Information | DMSO : 33.33 mg/mL (38.44 mM; Need ultrasonic) |
| SMILES | O=C(N[C@@H]1C(N[C@H](CC)C(N2[C@](CCC2)([H])C(N(C)[C@@H](CC3=CC=C(N(C)C)C=C3)C(N4[C@](CC(CC4)=O)([H])C(N[C@@H](C5=CC=CC=C5)C(O[C@@H]1C)=O)=O)=O)=O)=O)=O)C6=NC=CC=C6O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?