Learn More
Epredia™ PrintMate™ Biopsy Cassettes in Tubes, Aqua
Product Code. 15467574 Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1500
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15467574 Aqua Pack of 1500
15477574 Blue Pack of 1500
15497574 Green Pack of 1500
15407584 Gray Pack of 1500
15417584 Lilac Pack of 1500
15427584 Orange Pack of 1500
15437584 Pink Pack of 1500
15447584 Tan Pack of 1500
15457584 White Pack of 1500
15467584 Yellow Pack of 1500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15467574 Supplier Epredia™ Supplier No. A84210020
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. Packed in tubes, PrintMate Cassettes are intended for high throughput printing, and provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
PrintMate Biopsy Cassettes in pre-packed tubes. 20 tubes X 75 cassettes, total 1500 cassettes.
TRUSTED_SUSTAINABILITY
Specifications
PrintMate Compatible Yes
Color Aqua