missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRIMA1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15988194
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15988194 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15988194 Supplier Invitrogen™ Supplier No. PA565453

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KRKPLRKDENG TSVAEYPMSA SQSNKGVDVN NAVV Highest antigen sequence identity to the following orthologs - mouse 91%, rat 89%.

Required to anchor acetylcholinesterase (ACHE) to the basal lamina of the neuromuscular junction and to the membrane of neuronal synapses in brain. Also able to organize ACHE into tetramers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRIMA1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRIMA1
Gene Accession No. Q86XR5
Gene Alias acetylcholinesterase membrane anchor precursor PRiMA; PRIMA; PRIMA1; proline rich membrane anchor 1; proline-rich membrane anchor 1
Gene Symbols PRIMA1
Host Species Rabbit
Immunogen Recombinant Human PRIMA1. Recombinant protein control fragment (Product #RP-95709).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145270
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.