missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Presenilin-1 Antibody (APS 11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
213.00 €
Specifications
| Antigen | Presenilin-1 |
|---|---|
| Clone | APS 11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:1000, ELISA 4.5 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 0.75 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 0.75 μg/mL |
| Applications | Western Blot |
Description
Presenilin-1 Monoclonal specifically detects Presenilin-1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Presenilin-1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Protein S182, PS-1, PS1presenilin-1, PSNL1, S182 | |
| PSEN1 | |
| IgG1 | |
| Protein G purified | |
| No cross-reactivity is seen with presenilin 2. |
| APS 11 | |
| Western Blot 1:100 - 1:1000, ELISA 4.5 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 0.75 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 0.75 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| P49768 | |
| 5663 | |
| Synthetic peptide corresponding to residues C H(21) L S N T V R S Q N D N R E(34) of human PS1. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title