missing translation for 'onlineSavingsMsg'
Learn More

PRELID1 Monoclonal Antibody (OTI2A6), TrueMAB™, OriGene

Product Code. 17504654
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17504654 30 μL 30µL
17584644 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17504654 Supplier Origene Technologies Supplier No. TA811047S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRELID1 Monoclonal antibody specifically detects PRELID1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PRELID1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PCCA
Gene Accession No. P05165
Gene Alias 25 kDa protein of relevant evolutionary and lymphoid interest; 2610524G07Rik; CGI-106; Preli; PRELI domain containing 1; PRELI domain-containing protein 1, mitochondrial; PRELID1; protein of relevant evolutionary and lymphoid interest; PX19; PX19 homolog; px19-like protein; RGD1308082; SBBI12
Host Species Mouse
Immunogen Full length human recombinant protein of human PCCA produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5095
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.